Anti-Connexin 32/GJB1 Rabbit Polyclonal Antibody
Catalog # BSBTA01050
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Connexin 32/GJB1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Connexin 32/GJB1 detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: Connexin 32/GJB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat