Anti-14-3-3 sigma Rabbit Polyclonal Antibody
BSBTA01127
- Antibody type:Primary
- Antigen name:14-3-3 sigma
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for 14-3-3 sigma detection. Tested with WB, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: 14-3-3 sigma
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat