Anti-VEGF Receptor 1 Rabbit Polyclonal Antibody
Catalog # BSBTA00534-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:VEGF Receptor 1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for VEGF Receptor 1 detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: VEGF Receptor 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat