Anti-Prothrombin Rabbit Polyclonal Antibody
Catalog # BSBTA00044
Supplier: Boster Bio
Specifications
- Druh protilátky:Primární
- Název antigenu:Prothrombin
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Reaktivita:Human
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Prothrombin detection. Tested with WB, IHC-P, Direct ELISA in Human.
Type: Primary
Antigen: Prothrombin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human