Anti-Aquaporin 9 Rabbit Polyclonal Antibody
Catalog # BSBTA03638-1
Dodavatel: Boster Bio
Specifikace
- Antibody type:Primary
- Antigen name:Aquaporin 9
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Pk:100 µG
Specifikace
O tomto produktu
Rabbit IgG polyclonal antibody for Aquaporin 9 detection. Tested with WB in Human;Mouse;Rat.
Type: Primary
Antigen: Aquaporin 9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat