Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EPO Receptor Rabbit Polyclonal Antibody
Anti-EPO Receptor Rabbit Polyclonal Antibody
Catalog # BSBTA00427-2
Supplier:  Boster Bio
undefined
Anti-EPO Receptor Rabbit Polyclonal Antibody
Catalog # BSBTA00427-2
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EPO Receptor
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Rat,
    Mouse
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for EPO Receptor detection. Tested with WB, Direct ELISA in Mouse;Rat.

Type: Primary
Antigen: EPO Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat