

Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:H₃CCH₂OH
:46.07 g/mol
UN:1170
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (V/V):min. 97.2%
- Denatonium benzoate (ppm):12.3 - 14.7 ppm
- Methyl ethyl ketone (V/V):0.97 - 1.13 %
- 2-Propanol (V/V):0.97 - 1.13%
Specification Test Results
Frequently Bought Together