ContactUsLinkComponent
Certified reference material, soya-maize oil blend, BCR®
Certified reference material, soya-maize oil blend, BCR®
Catalog # PROOBCR-162R
CAS Number:  
Certified reference material, soya-maize oil blend, BCR®
Catalog # PROOBCR-162R
CAS Number:  
Specifications
  • Pack type:
    Ampoule, amber
  • Pk:
    5,5 g
  • Description:
    Soya-maize oil blend - fatty acids
Specifications
About this item
A unit contains approximately 5,5 gm soya-maize oil blend in 10 ml amber ampoule.
  • Fatty acid analysis
Certifications: IRMM certified.
Caution: Store in Freezer below –20 °C.