ContactUsLinkComponent
Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
 :  EUTECH
undefined
Pocket pH/°C testers, Thermo Scientific™ Eutech™ Elite pH
 :  EUTECH
  • Catalog No:
  • 662-2280
  • 662-2303
  • 662-2304
  • 662-2282
  • Model:
  • Standard
  • Standard
  • Spear
  • Spear
  • pH range:
  • −1,00...+15,00
  • −1,00...+15,00
  • −1,00...+15,00
  • −1,00...+15,00
  • pH resolution:
  • 0,01
  • 0,01
  • 0,01
  • 0,01
  • pH accuracy:
  • ±0,01 pH ±1 LSD
  • ±0,01 pH ±1 LSD
  • ±0,01 pH ±1 LSD
  • ±0,01 pH ±1 LSD
  • pH calibration:
  • Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18)
  • Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18)
  • Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18)
  • Up to 3 points (USA 4,01 / 7,00 / 10,01 or NIST 4,01 / 6,86 / 9,18)
  • Temperature range:
  • 0...+60,0 °C
  • 0...+60,0 °C
  • 0...+60,0 °C
  • 0...+60,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,5 + 1 LSD (0...+50)
    ±1,0 + 1 LSD (50...60)
  • ±0,5 + 1 LSD (0...+50 °C)
    ±1,0 + 1 LSD (50...60 °C)
  • -
  • -
  • Temperature compensation:
  • Automatic (ATC)
  • Automatic (ATC)
  • Manual (MTC)
  • Manual (MTC)
  • Display:
  • 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel)
  • 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel)
  • 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel)
  • 4-digit double line LCD with backlight (Graphic, dot matrix 80×100 pixel)
  • Power supply:
  • 4× AAA batteries
  • 4× AAA batteries
  • 4× AAA batteries
  • 4× AAA batteries
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • W×D×H:
  • 45×36×200 mm
  • 45×36×200 mm
  • 45×36×200 mm
  • 45×36×200 mm
  • Weight:
  • 0,165 kg
  • 0,165 kg
  • 0,165 kg
  • 0,165 kg
Replacement sensor