HC toxin
Supplier: Cayman Chemical
Certificates
About this item
HC Toxin is a cyclic tetrapeptide first isolated from H. carbonum (now C. carbonum), a pathogen of maize.
- Cell-permeable
- Reversible inhibitor of histone deacetylases (HDACs)
HC toxin has been shown to up-regulate the expression of 15-lipoxygenase-1 in colorectal cancer cells and induce fetal hemoglobin in human primary erythroid cells.
Specifications
- CAS No.:83209-65-8
- Form:A crystalline solid
- Protein synonyms:9S|Toxin I (Helminthosporium carbonum)|(6R|2-a][1,4,7,10]tetraazacyclododecine-1,4,7,10-tetranone|14aR)-3,6R-dimethyl-9S-(6-((S)-oxiran-2-yl)-6-oxohexyl)decahydropyrrolo[1
- Protein/peptide name:HC toxin
- Purity:>95%
- Molecular weight:436,5
- Molecular formula:C₂₁H₃₂N₄O₆
Specifications
Frequently Bought Together