Zinc standard solution
Catalog # PROONIST-3168A
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Matrix:10% HNO₃
- Pk:50 mL
- Description:NIST-3168A
- Element:Zinc
- Symbol:Zn
Specifications
About this item
This standard reference material is intended for use as a primary calibration standard for the quantitative determination of zinc.
A unit consists of 50 ml of a single-element solution in a high-density polyethylene bottle sealed in an aluminised bag.
Certifications: Traceable to NIST and SRM numbers.