Order Entry
Czech Republic
ContactUsLinkComponent
 
Chromium standard solution
  PROONIST-3112A
 :  
Chromium standard solution
  PROONIST-3112A
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃
  • Pk:
    5
  • Element:
    Chromium
  • Symbol:
    Cr

 

 

This reference standard solution is for use as a primary calibration standard for the quantitative determination of chromium.

One unit consists of five 10 ml sealed borosilicate glass ampoules of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of chromium.

 : Traceable to NIST and SRM numbers.