Organic reference standard, mussel tissue, NIST SRM 1974C
Katalogové číslo PROONIST-1974C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:5
- Standards:Reference standard
- Description:Mussel tissue
Specifications
About this item
This reference standard is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyl (PCB) congeners, chlorinated pesticides and polybrominated diphenyl ether (PBDE) congeners in marine bivalve mollusk tissue and similar matrices.
A unit consists of five jars each containing approximately 10 g (wet basis) of frozen tissue homogenate.
Certifications: Traceable to NIST and SRM numbers.