Order Entry
Czech Republic
ContactUsLinkComponent
Tris(2-chloroethyl)phosphate 100 μg/ml ≥98%
Tris(2-chloroethyl)phosphate 100 μg/ml ≥98%
Catalog # PROOCILULM93141.2
CAS Number:  
Tris(2-chloroethyl)phosphate 100 μg/ml ≥98%
Catalog # PROOCILULM93141.2
CAS Number:  

Specifications

  • Pk:
    1,2 mL
  • Seq:
    2080

Specifications

About this item

 :
 :285.49 g/mol
CAS Number:115-96-8
Formula:C₆H₁₂Cl₃O₄P
MDL Number:MFCD00000967