Order Entry
Czech Republic
ContactUsLinkComponent
 
Impurity and API standards, leflunomide impurity
 
Impurity and API standards, leflunomide impurity

 

  • CAS No.:
    163451-81-8
  • Formula:
    C₁₂H₉F₃N₂O₂
  • Description:
    Leflunomide Imp. B (EP): (2Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]but-2-enamide)

 

Frequently Bought Together