Order Entry
Czech Republic
ContactUsLinkComponent
Impurity and API standard, (2RS)-2-[(1,2-diphenyldiazanyl)carbonyl]hexanoic acid
Impurity and API standard, (2RS)-2-[(1,2-diphenyldiazanyl)carbonyl]hexanoic acid
  PROOMM0548.01
 :  
Impurity and API standard, (2RS)-2-[(1,2-diphenyldiazanyl)carbonyl]hexanoic acid
  PROOMM0548.01
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    3583-64-0
  • Description:
    (2RS)-2-[(1,2-diphenyldiazanyl)carbonyl]hexanoic acid
  • Formula:
    C₁₉H₂₂N₂O₃
  • Description:
    (2RS)-2-[(1,2-diphenyldiazanyl)carbonyl]hexanoic acid