Impurity and API standard, 6-cyclohexyl-4-methylpyridin-2(1H)-one
Catalog # PROOMM0418.03
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:67587-24-0
- Description:6-Cyclohexyl-4-methylpyridin-2(1H)-one
- Formula:C₁₂H₁₇NO
- Description:6-Cyclohexyl-4-methylpyridin-2(1H)-one