Impurity and API standard, (2S,5R,6R)-6-[[(2R)-2-[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylicacid
PROOMM0232.07
:
- Pk:100 mg
- Standards:Reference standards
- CAS No.:188112-75-6
- Description:(2S,5R,6R)-6-[[(2R)-2-[[(2R)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
- Formula:C₂₄H₂₆N₄O₇S
- Description:(2S,5R,6R)-6-[[(2R)-2-[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid