Order Entry
Czech Republic
ContactUsLinkComponent
Impurity and API standard, 5-chlorocarbonyliminostilbene
Impurity and API standard, 5-chlorocarbonyliminostilbene
  PROOMM0076.18
 :  
Impurity and API standard, 5-chlorocarbonyliminostilbene
  PROOMM0076.18
 :  

 

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    33948-22-0
  • Description:
    5H-dibenzo[b,f]azepine-5-carbonyl Chloride
  • Formula:
    C₁₅H₁₀ClNO
  • Description:
    5H-Dibenzo[b,f]azepine-5-carbonyl Chloride (5-Chlorocarbonyliminostilbene)