Impurity and API standard, N-[4-bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
PROOMM0064.02
:
- Pk:50 mg
- Standards:Reference standards
- CAS No.:41526-21-0
- Description:N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide
- Formula:C₁₄H₁₀BrClN₂O₂
- Description:N-[4-Bromo-2-(pyridin-2-ylcarbonyl)phenyl]-2-chloroacetamide