Order Entry
Czech Republic
ContactUsLinkComponent
Sulphur standard solutions for ICP, TraceCERT®, Supelco®
Sulphur standard solutions for ICP, TraceCERT®, Supelco®
Catalog # 18021-100ML
Dodavatel:  Merck
Číslo CAS:  
Sulphur standard solutions for ICP, TraceCERT®, Supelco®
Catalog # 18021-100ML
Dodavatel:  Merck
Číslo CAS:  

Specifikace

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ300
  • Element:
    Sulphur
  • Symbol:
    S

Specifikace

O tomto produktu

These standard solutions are used for ICP and AAS.

Certifications: Accordance with ISO/IEC 17025 and ISO 17034.