Organic reference standards, Meloxicam standard, BP
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
Specifications
- CAS No.:71125-38-7
- Formula:C₁₄H₁₃N₃O₄S₂
- Description:Meloxicam assay standard
Specifications
Frequently Bought Together