Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standards, pyridoxine hydrochloride assay standard, BP
Organic reference standards, pyridoxine hydrochloride assay standard, BP
  PROOBP458
 :  
Organic reference standards, pyridoxine hydrochloride assay standard, BP
  PROOBP458
 :  

 

  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    58-56-0
  • Description:
    Pyridoxine hydrochloride assay standard
  • Formula:
    C₈H₁₁NO₃·ClH
  • Description:
    Pyridoxine hydrochloride assay standard