Organic reference standards, cloxacillin benzathine assay standard, BP
Katalogové číslo PROOBP088
Dodavatel: LGC Standards PROMOCHEM
Číslo CAS:
Specifikace
- Pk:250 mg
- Standards:Reference Standard
- CAS No.:23736-58-5
- Description:Cloxacillin benzathine assay standard, BP088
- Formula:C₅₄H₅₆Cl₂N₈O₁₀S₂
- Description:Cloxacillin benzathine assay standard