Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standards, Cefalexin assay standard, BP
Organic reference standards, Cefalexin assay standard, BP
Catalog # PROOBP061
CAS Number:  
Organic reference standards, Cefalexin assay standard, BP
Catalog # PROOBP061
CAS Number:  

Specifications

  • Pk:
    150 mg
  • Standards:
    Reference standard
  • CAS No.:
    15686-71-2
  • Description:
    Cefalexin assay standard, BP-061
  • Formula:
    C₁₆H₁₇N₃O₄S
  • Description:
    Cefalexin assay standard

Specifications

About this item