Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standards, paclitaxel, Ph. Eur. RS
Organic reference standards, paclitaxel, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, paclitaxel, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    33069-62-4
  • Formula:
    C₄₇H₅₁NO₁₄
  • Description:
    Paclitaxel semi-synthetic for peak identification

Specifications

Frequently Bought Together