Order Entry
Czech Republic
ContactUsLinkComponent
N,N-Diethyl-p-phenylenediamine oxalate analytical reagent
N,N-Diethyl-p-phenylenediamine oxalate analytical reagent
Catalog # HACH608424
Supplier:  Hach
CAS Number:  
undefined
N,N-Diethyl-p-phenylenediamine oxalate analytical reagent
Catalog # HACH608424
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay:
    65 - 95%

Specification Test Results

About this item

 :
 :209.27 g/mol
CAS Number:62637-92-7
Formula:C₁₀H₁₆N₂·0.5 C₂H₂O₄
MDL Number:MFCD00070242