Organic reference standards, oxaliplatin, Ph. Eur. RS
: EDQM
- CAS No.:61825-94-3
- Formula:C₈H₁₄N₂O₄Pt
- Description:Oxaliplatin
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection