ContactUsLinkComponent
Standards for formaldehyde-2,4-dinitrophenyl-hydrazone, BCR®
Standards for formaldehyde-2,4-dinitrophenyl-hydrazone, BCR®
Catalog # PROOBCR-553-4
CAS Number:  
Standards for formaldehyde-2,4-dinitrophenyl-hydrazone, BCR®
Catalog # PROOBCR-553-4
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Formaldehyde 2,4-dinitrophenylhydrazone
Specifications
About this item
Used on glass fibre filters.
Ordering information: The kit consists of 2 samples of BCR-553 (spiked filter) and 1 sample of BCR-554 (blank filter).