Certified reference materials for environmental analysis, IRMM BCR®
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Diisopropyl ether stabilised for analysis
: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
Toluene, 99,5% for synthesis 1 * 5 L
: PanReac AppliChem
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
2-Propanol 99.5+%
: Thermo Fisher Scientific
Methanol, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
: Thermo Fisher Scientific