ContactUsLinkComponent
Oxygen demand, quality control standard
Oxygen demand, quality control standard
Katalogové číslo HACH2833510
Dodavatel:  Hach
Číslo CAS:  
undefined
Oxygen demand, quality control standard
Katalogové číslo HACH2833510
Dodavatel:  Hach
Číslo CAS:  
Specifikace
  • Concentration:
    396 mg/l as BOD
    613 mg/l as COD
    242 mg/l as TOC
  • Pk:
    1
  • Description:
    Oxygen quality control standard, 10 ml Voluette® ampoules
Specifikace
O tomto produktu
These mixed standards are ready to use, no dilutions are required. They contain BOD, COD and TOC.
Ordering information: Pack of 16×10 ml ampoules.