Ball valve lockouts, Perma-mount™
Supplier: Brady
Certificates
About this item
These retrofit devices stay permanently in place at the point of need. They ensure that critical valve energy isolation points are always ready for lockout.
- One-time installation is easy to complete in just a few minutes
- Permanently mounts onto valve body or pipe
- Eliminate the need to choose and carry the correct devices for each lockout event
- Securely retain the operating handle in the 90° fully-closed position
- Two sizes fit the full range of 1 to 10 cm diameter ball valves
- Designed to withstand a wide range of environmental conditions
Frequently Bought Together