Stainless steel unions
VICI Stainless Steel Unions are made for joining two pieces of ¹/₁₆" Ø tubing, or an ¹/₈" tube. They are precision machined with a focus on the concentricity of the bore as well as on the finish of the surfaces to give an absolute leak-free connection. These Unions have internal threads, which we recommend for most applications. Internal threads make stronger connections and internal fitting details provide the lowest dead volume for high performance instrumentation.
- Easy connection of solvent lines
- Low dead volume
: Unions are sold complete with all the necessary fittings. Nuts are SS303; ferrules are SS316; body is SS316.
Frequently Bought Together