Anti-IgG Goat Polyclonal Antibody (Biotin)
30-8067-M002
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Rabbit
- Epitope:Heavy and light chain
- Storage temperature:Store at 2-8 °C
- Pk:2 mg
Goat anti-Rabbit IgG (H&L) - Affinity Pure, Biotin Conjugate
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit