Order Entry
Czech Republic
ContactUsLinkComponent
Oversized Rayon OB/GYN Swabs
Oversized Rayon OB/GYN Swabs
Catalog # PMPC25-8081PR
Supplier:  PURITAN MEDICAL
Oversized Rayon OB/GYN Swabs
Catalog # PMPC25-8081PR
Supplier:  PURITAN MEDICAL

Specifications

  • Ø:
    12,7 mm (Tip) mm
  • Packed:
    10 boxes of 50 individually wrapped swabs
  • Sterile:
    +
  • Description:
    Oversized OB/GYN swabs
  • Length:
    201,6 mm
  • Pk:
    500

Specifications

About this item

Extra-absorbent oversized rayon tipped applicator with plastic handle.

  • Extra-absorbent large oversized tip
  • Long PS handles
  • Medical grade quality, sterile

Ideal for gathering specimens as well as OB/GYN preparation and examinations.

Packaging: 10 boxes of 50 wrapped packages.