ContactUsLinkComponent
Test kits, photometric, Tin, LCK cuvette tests
  705-3042
 :  Hach
 :  
Test kits, photometric, Tin, LCK cuvette tests
  705-3042
 :  Hach
 :  
  • Chemical method:
    Pyridinfluoron (PYF)
  • Pk:
    24 Tests
  • Description:
    Tin
  • Seq:
    278
  • Type:
    LCK359
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,1 - 2,0 mg/l Sn
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated)