Test kits, photometric, Sludge activity, LCK cuvette tests
Katalogové číslo HACHLCK318
Dodavatel: Hach
Číslo CAS:
Specifikace
- Chemical method:Colorimetric
- Pk:24 Tests
- Description:Sludge activity
- Seq:267
- Type:LCK318
- For unit:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Range:5 - 200 µg formazan (SA)
0 - 500 % Residual activity (RA)
Specifikace
O tomto produktu
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C
Certifications: DIN 38412-3