Test kits, photometric, BOD₅, LCK cuvette tests
Supplier: Hach
Certificates
About this item
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated)
Certifications: EN 1899-1