Water, BAKER INSTRA-ANALYZED® Plus, J.T.Baker®
9381.1000
: Avantor
:
- Pk:1 L
- Seq:5037
- Colour (Hazen):max. 5
- LC Gradient Suitability Test:passes test
- at 200 nm:max. 0.01
- at 254 nm:max. 0.005
- at 300 nm:max. 0.005
- at 400 nm:max. 0.005
- Fluorescence (as Quinine):max. 100 ppt
- Residue after Evaporation:max. 1 ppm
- Resistivity (at time of bottling):min. 18 megaohm
- Total Organic Carbon:max. 20 ppb
:
:18.02 g/mol
:7732-18-5
:H₂O
:MFCD00011332