Order Entry
Czech Republic
ContactUsLinkComponent
D-(+)-Sucrose, GenAR® NF, Macron Fine Chemicals™
D-(+)-Sucrose, GenAR® NF, Macron Fine Chemicals™
Catalog # 7724-88
CAS Number:  
undefined
D-(+)-Sucrose, GenAR® NF, Macron Fine Chemicals™
Catalog # 7724-88
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • GMP Manufactured Product:
  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • Meets N.F. Requirements:
  • Store in a tightly closed container.:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • NF - Identification:
    Passes Test
  • NF - Appearance of Solution:
    Passes Test
  • NF - Color Value:
    ≤45
  • NF - Conductivity:
    ≤35 µS/cm
  • NF - Optical Rotation (+):
    66.3 - 67.0 Degree
  • NF - Reducing Sugars:
    Passes Test
  • NF - Sulfite (SO2):
    ≤10 ppm
  • NF - Loss on Drying at 105 ºC:
    ≤0.1 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Color Value:
    ≤45
  • EP/BP - Conductivity, µS cm⁻¹:
    ≤35
  • EP/BP - Optical Rotation (+):
    66.3 - 67.0 Degree
  • EP/BP - Sulfite (SO₂):
    ≤10 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤0.1 %
  • Endotoxin Concentration (EU/g):
    ≤250
  • EP/BP - Reducing Sugars:
    Passes Test
  • Additional Tests - Dextrins:
    Passes Test
  • Additional Tests - Lead (Pb):
    ≤0.5 ppm

Specification Test Results

About this item

 :
 :342.3 g/mol
CAS Number:57-50-1
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626