Sodium sulphite, anhydrous ≥98.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 8064-20
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Case, Mall STAKMOR
- Pk:12 kg
- Seq:4187
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 98.0 %
- ACS - Assay (Na₂SO₃):≥98.0 %
- ACS - Chloride (Cl):≤0.02 %
- ACS - Free Acid:Passes Test
- ACS - Heavy Metals (as Pb):≤0.001 %
- ACS - Iron (Fe):≤0.001 %
- ACS - Titrable Free Base (meq/g):≤0.03
- ACS - Insoluble Matter:≤0.005 %
- Solubility:Passes Test
Specification Test Results
About this item
:
:126.04 g/mol
Merck Index:13,08757
Melting Pt:>500 °C
Density:2.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-821-4
REACH:01-2119537420-49
CAS Number:7757-83-7
Formula:Na₂SO₃
MDL Number:MFCD00003503