Order Entry
Czech Republic
ContactUsLinkComponent
 
Sodium sulphate, anhydrous ≥99.0% (dry basis) USP, Macron Fine Chemicals™
 
undefined
Sodium sulphate, anhydrous ≥99.0% (dry basis) USP, Macron Fine Chemicals™
 

 

  • Bulk Pharmaceutical Chemical:
    = 99.0 %
  • USP - Assay (Na₂SO₄) (dried basis):
    ≥ 99.0 %
  • USP - Acidity or Alkalinity (mL):
    ≤ 0.50
  • USP - Chloride (Cl):
    ≤ 0.005 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Matter:
    ≤ 0.01 %
  • USP - Loss on Drying at 105°C:
    ≤ 0.5 %
  • Thru U.S. No. 50 Sieve, For Information Only:
    = 0.5 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    For Information Only

 

 

Frequently Bought Together