Order Entry
Czech Republic
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    98.0 - 103.0%
  • Meets U.S.P Requirements:
    Passes Test
  • Meets B.P. Chemical Specifications:
    ≤8 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.014%
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • Appearance (White crystals or granules):
    Passes Test
  • USP - Assay (anhydrous basis):
    98.0 - 103.0 %
  • USP - Aluminum, Calcium, and Related Elements:
    Passes Test
  • USP - Arsenic (As):
    ≤ 8 ppm
  • USP - Chloride (Cl):
    ≤ 0.014 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.2 %
  • USP - pH (1 in 20):
    4.1 - 4.5
  • USP - Sulfate (SO₄):
    ≤ 0.15 %
  • USP - Water (H₂O):
    10.0 - 15.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Identification C:
    Passes Test
  • EP - Assay (anhydrous basis):
    98.0 - 100.5 %
  • EP - Chloride (Cl):
    ≤ 200 ppm
  • EP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP - Reducing Substances:
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • EP - Iron (Fe):
    ≤ 10 ppm
  • EP - Appearance of Solution:
    Passes Test
  • EP - pH:
    4.2 - 4.5
  • BP - Identification A:
    Passes Test
  • BP - Identification B:
    Passes Test
  • BP - Identification C:
    Passes Test
  • BP - Identification D:
    Passes Test
  • BP - Acidity:
    4.2 - 4.5
  • BP - Clarity and Color of Solution:
    Passes Test
  • BP - Arsenic (As):
    ≤ 2 ppm
  • BP - Iron (Fe):
    ≤ 10 ppm
  • BP - Chloride (Cl):
    ≤ 200 ppm
  • BP - Sulfate (SO₄):
    ≤ 300 ppm
  • BP - Reducing Substances:
    Passes Test
  • BP - Loss on Drying at 130°C:
    11.5 - 14.5 %
  • BP - Assay (dried basis):
    98.0 - 100.5 %
  • JPE - Assay (dried basis):
    ≥ 98.0 %
  • JPE - Identification:
    Passes Test
  • JPE - pH:
    4.1 - 4.7
  • JPE - Clarity and Color of Solution:
    Passes Test
  • JPE - Chloride (Cl):
    ≤ 0.005 %
  • JPE - Sulfate (SO4):
    ≤ 0.019 %
  • JPE - Heavy Metals:
    ≤ 10 ppm
  • JPE - Arsenic (As):
    ≤ 2 ppm

Specification Test Results

Related Information

Frequently Bought Together