

di-Sodium tetraborate decahydrate 99.0-105.0% NF, Macron Fine Chemicals™
Catalog # 7418-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Fibre drum
- Pk:300 lb
- Seq:7227
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:99.0 - 105.0 %
- NF - Assay (Na₂B₄O₇·10H₂O):99.0 - 105.0 %
- NF - Carbonate and Bicarbonate:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
Specification Test Results
About this item
:
:381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
EINECS:215-540-4
REACH:01-2119490790-32
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:1303-96-4
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193