Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets F.C.C. Requirements:
    Passes Test
  • Meets U.S.P Requirements:
    ≤0.015%
  • Food GMP Manufactured Product:
    ≤0.25%
  • GMP Manufactured Product:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤20 ppm
  • Bulk Food Chemical:
    ≤0.015%
  • USP - Identification:
    Passes Test
  • USP - Limit of Sulfur Compounds:
    ≤0.015 %
  • USP - Loss on Drying:
    ≤0.25 %
  • USP - Insoluble Substances:
    Passes Test
  • USP - Normal Carbonate:
    Passes Test
  • USP - Limit of Ammonia:
    ≤20 ppm
  • USP - Chloride (Cl):
    ≤0.015 %
  • USP - Arsenic (As):
    ≤2 ppm
  • USP - Assay (NaHCO₃) (dried basis):
    99.0 - 100.5 %
  • FCC - Identification:
    Passes Test
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Loss on Drying:
    ≤0.25 %
  • FCC - Assay (NaHCO₃) (dried basis):
    99.0 - 100.5 %
  • Appearance (White powder):
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together