Order Entry
Czech Republic
ContactUsLinkComponent
(±)-1,2-Propanediol ≥99.5% USP, Multi-Compendial, J.T.Baker®
(±)-1,2-Propanediol ≥99.5% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
(±)-1,2-Propanediol ≥99.5% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets U.S.P Requirements:
  • Meets J.P. Chemical Specifications:
  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • USP - Specific Gravity at 25°/25°C:
    1.035 - 1.037
  • USP - Acidity:
    Passes Test
  • USP - Water (H2O)(by Karl Fischer titrn):
    ≤ 0.20 %
  • USP - Identification A (IR):
    Passes Test
  • USP - Identification B - Diethylene Glycol:
    ≤ 0.10 %
  • USP - Identification B - Ethylene Glycol:
    ≤ 0.10 %
  • USP - Identification C:
    Passes Test
  • USP - Residue on Ignition:
    ≤ 0.007 %
  • USP - Chloride (Cl):
    ≤ 0.007 %
  • USP - Sulfate (SO₄):
    ≤ 0.006 %
  • USP - Assay (CH₃CHOHCH₂OH):
    ≥ 99.5 %
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • EP/BP - Relative Density (d²º₂₀):
    1.035 - 1.040
  • EP/BP - Refractive Index at 20°C:
    1.431 - 1.433
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Identification D:
    Passes Test
  • EP/BP - Appearance:
    Passes Test
  • EP/BP - Acidity:
    Passes Test
  • EP/BP - Oxidizing Substances:
    Passes Test
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Water (H₂O):
    ≤ 0.2 %
  • EP/BP - Ash (sulfated):
    ≤ 0.01 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Specific Gravity at 20°/20°C:
    1.035 - 1.040
  • JP - Acidity:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.007 %
  • JP - Sulfate (SO₄):
    ≤ 0.002 %
  • JP - Heavy Metals (as Pb):
    ≤ 5 ppm
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Glycerin:
    Passes Test
  • JP - Water (H₂O):
    ≤ 0.5 %
  • JP - Residue on Ignition:
    ≤ 0.005 %
  • JP - Distilling Range (Lower):
    184 - 189 °C
  • JP - Distilling Range (Upper):
    184 - 189 °C
  • JP - Ethylene Glycol:
    ≤ 0.1 %
  • JP - Diethylene Glycol:
    ≤ 0.1 %
  • JP - Any Other Impurity:
    ≤ 0.1 %
  • JP - Total Impurities:
    ≤ 1.0 %

Specification Test Results

Related Information

Frequently Bought Together