
Potassium thiocyanate ≥98.5% (Ag basis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:333-20-0
:
:KSCN
:MFCD00011413
:12,07866
:500 °C (1013 hPa)
:175 °C
:1.61 g/cm³ (20 °C)
:97.18 g/mol
:206-370-1
:GHS07
:Ambient
- Appearance:Passes Test
- Assay (KSCN) (by Ag titrn):≥98.5 %
- Insoluble in H₂O:≤0.005 %
- pH of 5% Solution at 25°C:5.3 - 8.7
- Chloride (Cl):≤0.005 %
- Sulfate (SO₄):≤0.005 %
- Ammonium (NH₄):≤0.003 %
- Sodium (Na):≤0.005 %
- Iodine Consuming Substances (meq/g):≤0.004
- Use Test:Passes Test
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤2.0 ppm
Frequently Bought Together