Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, ACS, J.T.Baker®
Catalog # 7390-19
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Potassium dihydrogen phosphate 98.0-100.5% (dried basis) NF, ACS, J.T.Baker®
Catalog # 7390-19
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    98.0 - 100.5 %
  • NF - Assay (KH₂PO₄) (dried basis):
    98.0 - 100.5 %
  • NF - Arsenic (As):
    ≤3 ppm
  • NF - Fluoride (F):
    ≤10 mg/kg
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Insoluble Substances:
    ≤0.2 %
  • NF - Lead (Pb):
    ≤5 ppm
  • NF - Loss on Drying at 105°C:
    ≤1.0 %
  • ACS - Assay (KH₂PO₄) (by acidimetry):
    ≥99.0 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤0.002 %
  • ACS - Loss on Drying at 105°C:
    ≤0.2 %
  • ACS - Nitrogen Compounds (as N):
    ≤0.001 %
  • ACS - pH of 5% Solution at 25°C:
    4.1 - 4.5
  • ACS - Sodium (Na):
    ≤0.005 %
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Solubility (10 in 100):
    Passes Test

Specification Test Results

About this item

 :
 :136.09 g/mol
 :GHS07
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401