Order Entry
Czech Republic
ContactUsLinkComponent
 
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
  5279.2500
 :  Avantor
   Safer Human and Environmental Health
 :  
undefined
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
  5279.2500
 :  Avantor
   Safer Human and Environmental Health
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

 

 

  • Assay (by GC):
    min. 99.9 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Water (KF):
    max. 0.02 %

 

 

 :
 :32.04 g/mol
 :1230
 :3,II
 :67-56-1
 :CH₃OH
 :MFCD00004595