Order Entry
Czech Republic
ContactUsLinkComponent
Mercury(II) chloride ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Toxic
Health
Environment
Mercury(II) chloride ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2594-01
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Toxic
Health
Environment
Mercury(II) chloride ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2594-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (HgCl₂):
    ≥99.5 %
  • Residue after Reduction:
    ≤0.020 %
  • Solution in Ether:
    Passes Test
  • Iron (Fe):
    ≤0.002 %

Specification Test Results

About this item

 :
 :271.5 g/mol
UN:1624
ADR:6.1,II
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:7487-94-7
Formula:HgCl₂
MDL Number:MFCD00011041